Cole-Parmer Essentials Optic Cleaning Wipes; 100/PK
Representative image only.
Cole-Parmer Essentials Item # WZ-86476-50
Made of lint-free polypropylene microfiber cloth
  • Clean eye glasses — removes oil, grease, and lint
  • Reclosable container
  • Packaged 100 per polyethylene canister
  • Each container is packaged for cleanroom use
  • 1 year warranty
£17.46GBP / Pkg of 100
£20.95GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Format30% isopropyl alchol and 70% deionized water
  • Width (InchShort)6
  • Height (in)7 1/2
  • Applicationseye glasses
  • DescriptionOptic Cleaning Wipes;
  • Warranty1 year
1 Year Warranty
Key Features
  • Clean eye glasses — removes oil, grease, and lint
  • Reclosable container
  • Packaged 100 per polyethylene canister
  • Each container is packaged for cleanroom use
  • 1 year warranty
More About this Item
These optic cleaning wipes are individually dispensed and comprise of 30% isopropyl alcohol and 70% deionized water, making them ideal for cleaning glasses. Ready to use in an instant, the 6" x 7 1/2" polypropylene, wood pulp, viscose blend cloth is soft, lint-free, and won't scratch your glasses.