This item is now discontinued. Please browse similar items
Cole-Parmer Mini-Gel System Comb, 8/16 Wells, 2.0 mm Thick; for 28559-10
Representative image only.
Cole-Parmer Item # WZ-28559-62
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Comb Thickness (mm)2
  • DescriptionMini-Gel System Comb, 8/16 Wells, 2.0 mm Thick; for 28559-10
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Combs are dual sided and height adjustable. Available in three thicknesses to accommodate different sample volumes.