This item is now discontinued. Please browse similar items
Cole-Parmer Mini-Gel System 1-Place Gel Caster Tray for 28559-10
Representative image only.
Cole-Parmer Item # WZ-28559-85
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMini-Gel System 1-Place Gel Caster Tray for 28559-10
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Gel Casters allow you to precast flat, even gels using the gel trays sold above. Casters accept one, three, or six trays. Casters feature a three-point leveling base and soft neoprene side gaskets that secure trays in place and effectively seal their ends during casting.