This item is now discontinued. Please browse similar items

Cole-Parmer Manual Slide Staining Stations-6 position

Representative image only.
Cole-Parmer Item # WZ-48578-74
Three-, six-, and twelve-well configurations are available to meet your staining needs
  • Built-in handles allow for convenient moving around the lab
  • Interlocking design allows multiple units to be attached together
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Number of slides)6
  • Max Temperature (° F)167
  • Max Temperature (° C)75
  • External Width (in)15
  • External Height (in)6
  • External Depth (in)6
  • External Width (cm)38.1
  • External Height (cm)15.24
  • External Depth (cm)15.24
  • DescriptionManual Slide Staining Stations-6 position
  • Warranty1 year
1 Year Warranty

Key Features

  • Built-in handles allow for convenient moving around the lab
  • Interlocking design allows multiple units to be attached together
  • 1 year warranty

More About this Item

Choose the well configuration that meets your application requirements. The metal-extruded frames interlock for back-to-back connections of multiple racks. Stations feature wells that are deep enough to immerse 1" x 3" slides. These plastic wells hold up to 250 mL of reagent, are solvent resistant, and include lids for limiting reagent evaporation. The three-position model has a removable plastic top that covers the entire assembly. Larger sizes feature a see-through, sliding plastic cover that allows access from either end, further reducing the chance of evaporation between staining protocols.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat