This item is now discontinued. Please browse similar items
Cole-Parmer Loop-Powered 2-Meter Enclosure, Plastic, NEMA 4X
Representative image only.
Cole-Parmer Item # WZ-30007-63
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DIN Size1/8\"
  • EnclosureNEMA 4X
  • DescriptionLoop-Powered 2-Meter Enclosure, Plastic, NEMA 4X
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All power comes directly from the 4 to 20 mA input signal—no additional AC power source is required! Explosion-proof models provide safe use in hazardous locations and are easy to install and calibrate. These rugged, yet economical meters are ideal for use in hostile environments. Choose the backlit display option when mounting the meters in poor lighting conditions. Meters and backlighting option are powered directly from the 4 to 20 mA loop; no need for an AC power source, batteries, or additional wiring. Models with explosion-proof enclosures meet specifications for use in Class I, groups B, C, D and Class II, for Groups E, F, and G locations.
Rugged housing handles tough industrial environments or select
an optional enclosure to keep your meters in a control area. Enclosures are available in a choice of three materials and can accommodate up to six loop-powered panel meters. Check individual details.