Cole-Parmer ISE Electrode; Calcium
Representative image only.
Cole-Parmer Item # WZ-27077-02
Double-junction electrodes easily connect to controller with stripped wire ends
  • Compatible with ISE controller 56105-00
  • 1 year warranty
PRODUCT OPTIONS
£316.74GBP / Each
£380.09GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Ion-Selective ElectrodeCalcium
  • ConnectorTinned ends
  • DescriptionISE Electrode; Calcium
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with ISE controller 56105-00
  • 1 year warranty
More About this Item
These industrial sealed double-junction electrodes easily connect to the ion selective controller. Industrial electrodes will withstand the conditions of your in-line application. Electrodes measure 12 mm dia x 80 mm L with 10-ft cord.