This item is now discontinued. Please browse similar items
Cole-Parmer Integrated Assembly High-pH Probe, SJ/100Ohm RTD; Plug/Tinned
Representative image only.
Cole-Parmer Item # WZ-27009-28
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Min pH (pH)0
  • Max pH (pH)14
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • ConnectorTinned ends
  • DescriptionIntegrated Assembly High-pH Probe, SJ/100Ohm RTD; Plug/Tinned
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
These electrodes are ideal for applications involving high-sodium concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Use for measurements from 0 to 14 pH. Electrodes withstand temperatures from 23 to 230ºF (-5 to 110ºC).
Note: 100Ω RTD ATC elements conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ thermistor ATC elements is a standard