This item is now discontinued. Please browse similar items
Cole-Parmer Green Neoprene Stoppers, Assorted Standard Sizes 00-8; 107/Pk
Representative image only.
Cole-Parmer Item # WZ-62991-90
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number00-8
  • ColorGreen
  • MaterialNeoprene
  • DescriptionGreen Neoprene Stoppers, Assorted Standard Sizes 00-8; 107/Pk
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.