Cole-Parmer Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.4 mm; 10/Pk
Representative image only.
Cole-Parmer Item # WZ-01998-56
Reusable one-piece design
  • Constructed of durable graphite
  • 1 month warranty
PRODUCT OPTIONS
£53.55GBP / Pkg of 10
£64.26GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGraphite Short Ferrules Compatible with Agilent Inlets, 1/16\" to 0.4 mm
  • Warranty1 month
1 Month Warranty
Key Features
  • Constructed of durable graphite
  • 1 month warranty
More About this Item
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450°C. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.