This item is now discontinued. Please browse similar items
Cole-Parmer Essentials Graduated Cylinder, PP 100 mL; 2/PK
Representative image only.
Cole-Parmer Essentials Item # WZ-06134-69
Don’t guess at volume
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Capacity (mL)100
  • Graduation Interval (mL)1
  • DescriptionGraduated Cylinder, PP 100 mL, 2/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
  • 1 year warranty
More About this Item
These polypropylene (PP) cylinders feature excellent transparency. Molded in one piece for enhanced durability. Cylinders have raised graduations for easy reading. Autoclavable.