This item is now discontinued. Please browse similar items
Cole-Parmer Gel Tray for High-Throughput Horizontal Gel System
Representative image only.
Cole-Parmer Item # WZ-28562-70
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGel Tray for High-Throughput Horizontal Gel System
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Gel Trays give you the flexibility of pouring gels in the unit using the tapered baffles or pouring gels outside of the unit by taping the ends of the tray. Trays are UV-transparent acrylic. Multiple comb slots let you cast gels for simultaneous short runs.
With 8 comb slots; pathlength of 2.5 cm/slot