Cole-Parmer Air Compressor for Compact Fluidized Sand Baths; 230 VAC
Representative image only.
Cole-Parmer Item # WZ-01184-83
  • 3 year warranty
  • Previous part number F119D
PRODUCT OPTIONS
£2,179.64GBP / Each
£2,615.57GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCompact fluidized sand baths 01184-00, 01184-02, and 01189-50
  • Power (VAC)230
  • Power (Hz)50
  • Power (watts)400
  • DescriptionAir Compressor for Compact Fluidized Sand Baths; 230 VAC
  • Warranty3 Years
3 Year WarrantyConformite Europeenne
Key Features
  • 3 year warranty
  • Previous part number F119D
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.