Cole-Parmer Fermentation DO Probe; SS/Polarographic/200 mm, 19 mm
Representative image only.
Cole-Parmer Item # WZ-05726-22
Designed for fermentation applications where probes are repeatedly steam-sterilized
  • 316 SS Construction
  • Polarographic
  • 1 year warranty
PRODUCT OPTIONS
£1,019.63GBP / Each
£1,223.56GBP (incl VAT)
Include Probe Cables…
Cole-Parmer® DO Probe Cables
£88.52 - £117.15GBP / Each
£106.22 - £140.58GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes and Sensors
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Min Temperature (° F)32
  • Max Temperature (° F)176
  • Temperature compensationAutomatic
  • Length (cm)20
  • Diameter (cm)1.9
  • DescriptionFermentation DO Probe; SS/Polarographic/200 mm, 19 mm
  • Warranty1 year
1 Year Warranty
Key Features
  • 316 SS Construction
  • Polarographic
  • 1 year warranty
More About this Item
All models feature a polarographic sensor, and are available in a variety of body diameters, mounting styles, and immersion lengths. Temperature compensated probes feature 100 Ω RTD temperature measuring sensors. Order cables separately.