This item is now discontinued. Please browse similar items
Cole-Parmer Fermentation 1 or 2 Liter System Antifoam Probe; 18mm Thread/100mm Immersion Depth
Representative image only.
Cole-Parmer Item # WZ-01919-60
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFermentation 1 or 2 Liter System Antifoam Probe; 18mm Thread/100mm Immersion Depth
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Order a probe according to your vessel size; all probes require cable 01919-65.