This item is now discontinued. Please browse similar items
Cole-Parmer Black Vinyl ESD Bench Height Ergonomic Chair In Chrome With Casters
Representative image only.
Cole-Parmer Item # WZ-90003-55
Static control chairs that make a difference
  • Electrostatic discharge materials to reduce static electricity
  • ESD non-marring bell glides or rolling casters suitable for hard and carpeted floors
  • The six-way adjustable arm rests fit all possible ergonomic seating configurations
  • Adjustable arm rests move up and down, in and out, and twist toward or away from the user
  • Highly durable molded polyurethane foam cushioning covered with static-dissipating, heavy-duty vinyl is easily cleaned
  • Available in black or blue seat/back color, with or without arm rests, and chrome foot rings
  • Warranty 15 years on parts, 5 years on upholstery, lifetime on pneumatic lift
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")25
  • Dimension22-29\" adjustable height (measured from the top of the seat to the floor)
  • MaterialVinyl
  • ColorBlack
  • DescriptionBlack Vinyl ESD Bench Height Ergonomic Chair in Chrome with casters
  • WarrantyWarranty 15 years on parts, 5 years on upholstery, lifetime on pneumatic lift
Key Features
  • Electrostatic discharge materials to reduce static electricity
  • ESD non-marring bell glides or rolling casters suitable for hard and carpeted floors
  • The six-way adjustable arm rests fit all possible ergonomic seating configurations
  • Adjustable arm rests move up and down, in and out, and twist toward or away from the user
  • Highly durable molded polyurethane foam cushioning covered with static-dissipating, heavy-duty vinyl is easily cleaned
  • Available in black or blue seat/back color, with or without arm rests, and chrome foot rings
  • Warranty 15 years on parts, 5 years on upholstery, lifetime on pneumatic lift
More About this Item
Polyurethane chairs have a seat tilt of eight degrees forward and three degrees backward. The seat tilt locks into place. With a curved back and tilting, and an articulating seat, the chair offers excellent support and maximum comfort. The dissipative static control materials meet ANSI/ESD-STM-12.