This item is now discontinued. We recommend the following as suggested replacements: WZ-24501-29
Cole-Parmer Electronic Pipette, Single Channel, 1 to 10 mL
Representative image only.
Cole-Parmer Item # WZ-24500-06
Improve efficiency by saving routine dispense programs for quick access
  • Program up to six presets with function, volume, speed, and number of dispenses
  • Nine pipetting functions cover common and specialized pipetting applications
  • Adjust the large color display's brightness and background to suit any lighting conditions
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableTip cone
  • Max Volume (µL)1000
  • Min Volume (µL)10000
  • Min Volume (mL)1
  • Max Volume (mL)10
  • Increments (µL)10
  • Accuracy (%)±0.4
  • Reproducibility≤ 0.2
  • Number Of Channels1
  • Tip EjectorYes, soft touch
  • BatteryRechargeable battery (included), charge via; USB (cable included) or AC (adapters included)
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Key Features
  • Program up to six presets with function, volume, speed, and number of dispenses
  • Nine pipetting functions cover common and specialized pipetting applications
  • Adjust the large color display's brightness and background to suit any lighting conditions
  • 1 year warranty
More About this Item
This electronic pipettor is easy to use with simple plain language menus reaching only three levels in depth for quick navigation. Straightforward procedure makes calibration in the lab a snap. Pipetting functions include manual, automatic, reverse, stepped, mixing, stepped + sequence, pipette + mix, aspirate, and gel loading.
Display shows selected function, volume, piston position, selected speed, counter cycle, and battery status.