Cole-Parmer Electrode cable, detachable, 10-ft, BNC
Representative image only.
Cole-Parmer Item # WZ-05662-56
For use with flow-through pH cells
  • 3 ft or 10 ft length
  • 1 year warranty
PRODUCT OPTIONS
£79.38GBP / Each
£95.26GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)120
  • Length (cm)300
  • DescriptionElectrode cable, detachable, 10-ft, BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • 3 ft or 10 ft length
  • 1 year warranty
More About this Item
Detachable cable features BNC connection for compatibility with most pH meters and controllers.