This item is now discontinued. We recommend the following as suggested replacements: WZ-04807-80
Cole-Parmer Digital Hot Plate, 12" x 12", metal plate, 230 VAC
Representative image only.
Cole-Parmer Item # WZ-04805-44
Get more accurate heat control
  • Digital setting and control of plate temperature
  • Be safe—flashing light warns when top plate is hot, even if the power is off
  • Push buttons and dial setting for fast temperature changes and simple control
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)230
  • Power (Hz)50/60
  • Power (watts)600
  • DescriptionDigital Hot Plate, 12\" x 12\", metal plate, 230 VAC
  • Warranty3 Years
3 Year Warranty
Key Features
  • Digital setting and control of plate temperature
  • Be safe—flashing light warns when top plate is hot, even if the power is off
  • Push buttons and dial setting for fast temperature changes and simple control
  • 3 year warranty
More About this Item
The coated aluminum top plate offers excellent heat transmission and has been combined with a state-of-the-art digital temperature control for rapid heating across the entire plate. The angled housing provides stability while deflecting spills away from the controls. The "Hot" warning light will flash whenever the plate temperature is above 50°C. An independent safety circuit protects against overheating and internal electronic components are protected against corrosion. Units accommodate multiple vessels, one large container, or even microscope slides.