Cole-Parmer Detachable Fermentation pH Electrode Cable; BNC, 5 ft
Representative image only.
Cole-Parmer Item # WZ-05661-08
Easily detachable electrode cable
  • Optimize performance electrodes with optional cables
  • 1 year warranty
PRODUCT OPTIONS
£69.86GBP / Each
£83.83GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ConnectorBNC
  • Cable Length (m)1.5
  • DescriptionDetachable Fermentation pH Electrode Cable; BNC, 5 ft
  • Warranty1 year
1 Year Warranty
Key Features
  • Optimize performance electrodes with optional cables
  • 1 year warranty
More About this Item
Coose from BNC or stripped wire cables. Stripped end cables are used to connect the electrodes to meters, controllers and transmeters that accept bare wire connections. Stripped end cables are not for temperature compensated use.
BNC cables are used to connect the electrodes to meters, controllers and transmeters that accept BNC connections (most common).