Cole-Parmer Essentials Cylindrical Closed-Dome Tank, HDPE, 30 Gal.
Representative image only.
Cole-Parmer Essentials Item # WZ-06318-70
FDA compliant
  • 1 year warranty
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • Wall thickness (in)0.187
  • Volume (Gal)30
  • Volume (Liters)114
  • Height (in)36 3/4
  • Height (cm)93.345
  • Outside Diameter (OD) (in)18
  • DescriptionCylindrical Closed-Dome Tank, HDPE, 30 Gal.
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All tanks have a threaded screw closure with gasket that secuely fits the 6 1/4" fill well. Two mounting flats near the bottom, set 180°> apart, permit installation of a bulkhead fitting up to 2". Domed bottom drains fluid into narrow trough around base rim. High-density polyethylene tanks have a 2" bung with with cap on the top that permits access for tubing or piping. All tanks are FDA compliant.
Thermo Scientific Nalgene-Reg TM Nalge Thermo Scientific Nunc International