Cole-Parmer CP 20 Vacuum Pump Oil, 58 cSt, 1 gal
Representative image only.
Cole-Parmer Item # WZ-79755-22
Ensures optimum pump performance
  • Resists oxidation and thermal degradation
PRODUCT OPTIONS
£202.60GBP / Gallon of 1
£243.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • FormLiquid
  • Viscosity (cSt)58
  • Volume (mL)3785
  • Volume (Qt)4
  • Volume (Gal)1
  • Vapor Pressure (torr)0.000003 at 25°C
  • Packaging3785 mL
  • DescriptionVacuum Pump Oil, 58 cSt, 1 gal
Key Features
  • Resists oxidation and thermal degradation
More About this Item
Vacuum pump oil is formulated to ensure optimum pump performance and pump life.
CP 20 exhibits a low vapor pressure and resists oxidation and thermal degradation—ideal for mechanical vacuum pumps. All types have a specific gravity of 0.86 g/mL.
The CP-20 diffusion pump oil is a double-distilled, highly refined hydrocarbon based diffusion pump fluid that offers high resistance to corrosion. This oil is recommended for processes such as plasma etching, LPCVP, and metallization applications. CP-20 offers an additional benefit to be an excellent quality alternative oil for mechanical pumps as well.
Discounts: Save 10% on 12-23 quarts, 15% on 24-35 quarts and 20% on 36 or more quarts. Save 10% on 4-11 gallons, 15% on 12-23 gallons and 20% on 24 or more gallons.