Cole-Parmer Conductivity Cell; SS K=1, PT100
Representative image only.
Cole-Parmer Item # WZ-35820-30
Economical replacements for a variety of controllers
  • Ten-foot cables with stripped ends
  • Flow-through, in-line, swage, and dip styles available
PRODUCT OPTIONS
£320.01GBP / Each
£384.01GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK-1
  • Body MaterialStainless Steel
  • DescriptionConductivity Cell; SS K=1, PT100
Key Features
  • Ten-foot cables with stripped ends
  • Flow-through, in-line, swage, and dip styles available
More About this Item
Cells withstand pressures up to 150 psi (at 135°C for SS; 25°C for CPVC and PVDF).