This item is now discontinued. Please browse similar items
Cole-Parmer Compact Illuminated Microscope, 30X; 1/Ea
Representative image only.
Cole-Parmer Item # WZ-98766-60
See in true color with high contrast and a sharply defined focus
  • Adjustable LED light source directs onto the exact area being examined
  • Choose the 98766-59 with snap-on slide attachment—view microscope slides anywhere!
SPECS & DESCRIPTION
Specifications & Description
  • Magnification30x
  • LensSingle
  • Mounting TypeHandheld
  • Light SourceLED
  • BatteryAA Battery (2)
  • ColorBlack
  • DescriptionCompact Illuminated Microscope, 30X; 1/Ea
1 Year Warranty
Includes
vinyl case and batteries.
Key Features
  • Adjustable LED light source directs onto the exact area being examined
  • Choose the 98766-59 with snap-on slide attachment—view microscope slides anywhere!
More About this Item
These handheld compact microscopes are perfect for inspection, quality control, clinical observation, cloth/paper examination, and plant studies. Case measures 1-1/2"W x 5-1/2"H x 7/8"D (3.8 x 14 x 2.2 cm) and is chemical- and shock-resistant ABS plastic—perfect for field use.