This item is now discontinued. Please browse similar items
Cole-Parmer Comfort Grip Tweezer, 5, 1/pk
Representative image only.
Cole-Parmer Item # WZ-07288-21
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 3/4
  • Overall Length (cm)10.07
  • MaterialStainless Steel
  • SterileNo
  • DescriptionComfort Grip Tweezer, 5, 1/pk
More About this Item
Comfort grips tweezers provide exceptional relief from hand fatigue. Tweezer tips are manufactured with stainless steel and can withstand temperatures to 190°C (375°F).