Cole-Parmer Combination pH Electrode, Glass, Double-Junction, BNC
Representative image only.
Cole-Parmer Item # WZ-59001-36
Faster response and increased stability across a pH range of 0 to 14
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 1 year warranty
PRODUCT OPTIONS
£195.25GBP / Each
£234.30GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRugged for TRIS, sulfates, proteins
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionCombination pH Electrode, Glass, Double-Junction, BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 1 year warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.