Cole-Parmer Essentials Clean Wipes, 100% DI Water; 100 Wipes/Canister
Representative image only.
Cole-Parmer Essentials Item # WZ-33675-05
Strong, nonwoven cloth is soft and lint-free – will not scratch surfaces
  • No more struggling with solutions and separate towels—ready to use
  • Individually sealed containers for use in cleanrooms
  • 1 year warranty
PRODUCT OPTIONS
£18.00GBP / Each
£21.60GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene, Wood Pulp, Viscose Blend
  • Width (in)6
  • Length (in)7 1/2
  • DescriptionClean-Wipes, 100% DI Water: 100 Wipes/Canister
  • Warranty1 year
1 Year Warranty
Key Features
  • No more struggling with solutions and separate towels—ready to use
  • Individually sealed containers for use in cleanrooms
  • 1 year warranty
More About this Item
Premoistened wipes are available either with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reagent-grade deionized water, or 100% pure reagent-grade deionized water. Individually dispensed wipes are handy and ready for instant use. Snap-on sealed top eliminates contamination from dust or dirt. Each canister is provided in shrink-wrap or a sealed bag to maintain cleanliness for use in clean rooms. Wipes are 6 x 7.5 inches and packaged 100 per polyethylene canister.