Cole-Parmer Cable, 4-pin to BNC w/ATC wires for 29045 series pH electrodes, 3-ft
Representative image only.
Cole-Parmer Item # WZ-29045-50
New glass formulation provides unparalled performance!
  • Lightening fast response time with superior accuracy and repeatabilty
  • Perfect for low-ionic strength solutions
  • Provides confident values over the entire pH range from 0 to 14
  • 1 year warranty
PRODUCT OPTIONS
£78.97GBP / Each
£94.76GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Temperature CompensationYes
  • ATC ConnectorTinned ends
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionCable, 4-pin to BNC w/ATC wires for 29045 series pH electrodes, 3-ft
  • Warranty1 year
1 Year Warranty
Key Features
  • Lightening fast response time with superior accuracy and repeatabilty
  • Perfect for low-ionic strength solutions
  • Provides confident values over the entire pH range from 0 to 14
  • 1 year warranty
More About this Item
Cole-Parmer's fast-response fermentation pH electrodes are made of a proprietary new formulation of low-impedance glass replaces all other formulas — allows you to streamlined inventories and eliminate wasted time trying to select the best electrode for your application. Use in low-ionic strength solutions, high pH testings where sodium error could be a problem, and at high temperatures up to 266°F (130°C). pH is registered in less than 20 seconds, with a typically accuracy of 0.02 pH units (meter dependant).
Chemical-resistant Ryton ''PPS'' PG13.5 threaded pressure fitting, PTFE spacer, and silicone o-ring ensure a leakproof connection to your assembly.
All electrodes feature double-junction reference for maximum lifespan.
Order detachable cap