Sherwood Scientific Butane primary regulator, "gaz" type cylinder
Representative image only.
Sherwood Scientific Mfr # 00108732 – Item # WZ-02655-24
Provides controlled presssure of gas
  • 1 year warranty
PRODUCT OPTIONS
£347.94GBP / Each
£417.53GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionButane primary regulator, \"gaz\" type cylinder
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A primary regulator is required to adjust the flow rate of fuel gas entering the flame photometer. Choose a regulator from the table according to your fuel type and cylinder valve outlet.
Note: All fuels should be free of heavy hydrocarbon deposits and regulated at the cylinder to 30 psi.