Cole-Parmer Beaker Positioning Cover for 0.75 gal and SS 1 gal Cleaners
Representative image only.
Cole-Parmer Item # WZ-08891-50
Achieve optimal cleaning results
  • Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers
  • 2 year warranty
PRODUCT OPTIONS
£61.98GBP / Each
£74.38GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Material304 SS
  • DescriptionBeaker Positioning Cover for 0.75 gal and SS 1 gal Cleaners
  • Warranty2 Years
Two year warranty
Key Features
  • Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers
  • 2 year warranty
More About this Item
Use polypropylene beaker positioning covers with beakers to clean small objects or for indirect cleaning. Cover suspends beakers above tank bottom for increased ultrasonic activity. Covers come with beaker suspension ring. Order beakers separately.
Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners. Use an insert tray or beaker positioning cover and beaker with these ultrasonic cleaners. Ultrasonic acticity is greatest starting about an inch from the bottom of the tank.