This item is now discontinued. Please browse similar items
Cole-Parmer B3D5000 3D rocker with flat mat; 100-240V, US Plug
Representative image only.
Cole-Parmer Item # WZ-04304-41
Undulating rocker for molecular applications
  • Easily adjust tilt angle from 0° to 10° without tools
  • Ideal for refrigerated or incubated environments
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description3D rocker with flat mat; 100-240V, US Plug
  • Warranty2 Years
Two year warranty
Key Features
  • Easily adjust tilt angle from 0° to 10° without tools
  • Ideal for refrigerated or incubated environments
  • 2 year warranty
More About this Item
This digital rocker is a versatile workhorse, providing gentle slow-moving waves to more vigorous rocking and circular rotation. Ideal for generating the three-dimensional motion commonly preferred for molecular biology applications. Easily adjust the time of operation and speed on the large user-friendly LCD control panel.