Cole-Parmer Autoclavable pH Probe, Refillable/DJ/Glass; 420 mm
Representative image only.
Cole-Parmer Item # WZ-29041-06
Double-junction electrodes
  • Sealed, gel-filled, double-junction reference electrodes require no refilling
  • Offers long-term stability
  • Extends service life
  • 1 year warranty
PRODUCT OPTIONS
£361.86GBP / Each
£434.23GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsAutoclavable, refillable
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Max Temperature (° C)130
  • DescriptionAutoclavable pH Probe, Refillable/DJ/Glass; 420 mm
  • Warranty1 year
1 Year Warranty
Key Features
  • Sealed, gel-filled, double-junction reference electrodes require no refilling
  • Offers long-term stability
  • Extends service life
  • 1 year warranty
More About this Item
These electrodes are designed for insertion in RADIOMETER mountings or Ingold nonpressurized assemblies 761-351, 761-31, 761-361, and 361CH-31 (12 mm diameter). Annular ceramic junction eliminates clogging problems and provides excellent stability. Operating temperature range is 23 to 266°>F (-5 to 130°>C). Order detachable cable separately to autoclave your glass-bodied electrode and vessel as a single unit -- simply detach the cable and place a protective cap over the cable connector before sterilizing.
Length listed in table refers to probe immersion length, which is equal to length plus 70 mm (except Ingold 761-31, where immersion length equals length plus 50 mm).