Environmental Express APC3535 Clear VOA Vials, 40 mL, Closed-Top Cap, Economy Case; 144/Cs
Representative image only.
Environmental Express Item # WZ-35207-25
Borosilicate glass vials meet EPA guidelines for contamination-free containers
  • Choose between 40- and 60-mL capacities
  • Include standard screw-top caps
  • Model number APC3535
PRODUCT OPTIONS
£69.86GBP / Case of 66
£83.83GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)40
  • Tube MaterialBorosilicate glass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypePTFE-lined screw-top
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Thread24 mm
  • SterileNo
  • AutoclavableNo
  • Level3
  • Special Features
  • DescriptionClear VOA Vials, 40 mL, Closed-Top Cap, Economy Case; 144/Cs
Key Features
  • Choose between 40- and 60-mL capacities
  • Include standard screw-top caps
  • Model number APC3535
More About this Item
Level 3 VOA vials are not certified and ideal for general use containers that meet EPA recommended guidlines. Vials come with closed-top caps with PTFE liner. Choose from clear vials to easily view samples or amber vials to protect light-sensitive samples.