Cole-Parmer ADCF Male Luer to 1/8" L Barb Adapter, PP, 25/pk
Representative image only.
Cole-Parmer Item # WZ-30800-24
£11.67GBP / Pkg of 25
£14.00GBP (incl VAT)
Cole-Parmer Stopcocks with Luer Connections; 3-way; male lock, Non-sterile
£16.65GBP / Pkg of 10
£19.98GBP (incl VAT)
Complete the connection
Cole-Parmer ADCF Female Luer to 1/8
£9.55GBP / Pkg of 25
£11.46GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 2Hose barb
  • Barb Size-Port 2 (in)1/8
  • DescriptionADCF Male Luer to 1/8" L Barb Adapter, PP, 25/pk
Certificate of Compliance
More About this Item
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving). Sold in packs of 25.
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
4 Questions | 4 Answers
Displaying questions 1-4
  • Q:
    Is this material not autoclaveable? Do you have an alternative part number that is the same, but autoclaveable?
    Asked on 11/18/2015 by Holly
    1 answer
    • STAFF EXPERT
      A:
      The 45505-04 and 45513-09 are the same size but made of different, autoclavable materials.
      Answered on 11/18/2015 by Polo from Technical Support
  • Q:
    Are luer lock fittings rated for vacuum? or pressure?
    Asked on 12/26/2014 by Al from MN
    1 answer
    • STAFF EXPERT
      A:
      I apologize, but they are not specifically rated for vacuum or pressure.
      Answered on 12/29/2014 by Anthony from Technical Support
  • Q:
    What is the difference between an "L" or "J" barb adapter?
    Asked on 12/17/2014 by Dawn
    1 answer
    • STAFF EXPERT
      A:
      The "J" and "L" do not designate anything. They were mistakenly included in the product description. Please ignore those. I apologize for the inconvenience.
      Answered on 12/17/2014 by Anonymous
  • Q:
    What would you use with this Male Luer Adapter?
    Asked on 8/15/2013 by Al from San Diego, Ca.
    1 answer
    • STAFF EXPERT
      A:
      This is designed to connect to a 1/8" ID flexible hose, and on the other end, a female luer connector.
      Answered on 8/15/2013 by Jack from Technical Support
Displaying questions 1-4
Do you have a question about this product? 