Cole-Parmer Essentials Cell and Tissue Culture Plates with Lid, Sterile, Polystyrene, Treated, 6-Well; 100/CS
Representative image only.
Cole-Parmer Essentials Item # WZ-01959-40
Single direction lid reduces the risk of cross-contamination
  • Gamma irradiation sterilized and individually packaged to prevent contamination
  • Lid stacking ring offers stability when handling tall stacks of plates
  • Alphanumeric markings provide easy identification of individual wells
  • Can safely withstand -20 to 70 °C
  • Features treated surface that provides a hydrophilic surface ideal for cell attachment
PRODUCT OPTIONS
£218.25GBP / Case of 100
£261.90GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells6
  • Min Working Well Volume (mL)2
  • Max Working Well Volume (mL)8
  • Well ShapeFlat
  • Culture Area (cm²)9.6
  • Surface TreatmentYes
  • Lid (Included)Yes
  • SterileYes
  • DescriptionCell and Tissue Culture Plates with Lid, Sterile, Polystyrene, Treated, 6-Well; 100/CS
Sterile
Includes
Lids
Key Features
  • Gamma irradiation sterilized and individually packaged to prevent contamination
  • Lid stacking ring offers stability when handling tall stacks of plates
  • Alphanumeric markings provide easy identification of individual wells
  • Can safely withstand -20 to 70 °C
  • Features treated surface that provides a hydrophilic surface ideal for cell attachment
More About this Item
Featuring a flat well bottom, these nonpyrogenic cell and tissue culture plates are ideal for cell growth and cell yields. The single direction lid reduces the risk of cross-contamination while a raised rim on each well nests into the lids rings to reduce evaporation. Lids have a stacking feature for added stability when handling tall stacks of plates. They are labeled with alphanumeric markings for easy identification of individual wells and can safely withstand temperatures from -20 to 70 °C.
Each well plate is constructed of clear polystyrene, is RNase/DNase free, sterilized by gamma irradiation (SAL 10-6), and packaged individually. Select a treated surface culture plate that provides a hydrophilic surface when cell attachment is desired, or an untreated surface culture plate when working with suspensions.