This item is now discontinued. Please browse similar items
Cole-Parmer 4-Place Electrophoresis System w/Casting Kit & Cooling; 10x1.5mm Well
Representative image only.
Cole-Parmer Item # WZ-28571-34
Increase throughput of your electrophoresis system – run up to four gels or four blots simultaneously in one unit!
  • Versatile unit accepts slab gels or Western blots
  • Designed for use with fast and efficient precast gels or traditional handcasting
  • Use with precast gels (see Related Product ad) to increase your throughput
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Solution Required (mL)1600
  • Gel Thickness (mm)1.5
  • Glass Width (cm)10
  • Number of Wells10
  • Description4-Place Electrophoresis System w/Casting Kit & Cooling; 10x1.5mm Well
  • Warranty3 Years
3 Year WarrantyConformite Europeenne
Key Features
  • Versatile unit accepts slab gels or Western blots
  • Designed for use with fast and efficient precast gels or traditional handcasting
  • Use with precast gels (see Related Product ad) to increase your throughput
  • 3 year warranty
More About this Item
These multipurpose gel systems are designed for use with either precast gels or traditional handcast gels to suit your needs. Sturdy design has no loose parts to keep track of and allows for easy loading and removal of your samples. Hinged-core design allows you to effortlessly extract the cassettes without removing the core from the tank—for quick sample processing and immediate reuse of buffers for your next set of samples. The color-coded anode and cathode safety features help with identification. Tank design includes a stir bar corral and hinged core.
Eliminate having to use several systems for your electrophoresis needs—this one unit lets you run up to four gels or four blots simultaneously! Unit uses 1.6 liters of buffer for either application. These systems accommodate 10 x 10 cm, 10 x 9 cm, or 10 x 8 cm precast gels. Available with or without the cooling system; use the cooling system for temperature controlled runs.
System with internal cooling includes: lower buffer reservoir with double-sided cooling labyrinth and tubing adapters, two cores to accommodate a total of four gels or four blotting cassettes, four blotting cassettes with sponge pads, adapter plate for running single gels, and safety cover with attached power leads.
Casting kit adds on: four sets of glass plates, four combs, four sets of spacers, four sets of gel wrap gaskets, and casting clamps.