This item is now discontinued. Please browse similar items
Cole-Parmer 332-3431-CP Waste System, 83 mm Cap, 4 x 1/16", 4 x 1/4" OD Tube Fittings, Exhaust Filter; 10 L
Representative image only.
Cole-Parmer Item # WZ-06064-12
Protect workers from inhaling dangerous fumes while collecting HPLC waste
  • Capture potentially dangerous solvent vapors
  • Safely collect waste with the high-density polyethylene (HDPE) carboy which features superior chemical compatibility
  • No need to remove tubing from the open cap to install a closed cap
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)10
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size83 mm
  • DescriptionCarboy Solvent Waste System, 83 mm Cap, 4 x 1/16\", 4 x 1/4\" OD Tube Fittings, Exhaust Filter; 10 L
Key Features
  • Capture potentially dangerous solvent vapors
  • Safely collect waste with the high-density polyethylene (HDPE) carboy which features superior chemical compatibility
  • No need to remove tubing from the open cap to install a closed cap
More About this Item
This heavy-duty solvent waste system is designed to collect liquid waste disposal from one or more HPLC systems while a carbon exhaust filter captures potentially harmful vapors emitted. The closed system helps eliminate spills and provides a safe and efficient way of containing, transporting, and disposing of aggressive organic solvents.
The polypropylene cap (PP) offers a wide range of tubing port configurations. The 10-L carboys come with 83 mm caps.