This item is now discontinued. Please browse similar items
Cole-Parmer 205-7002-CP Carboy Cap, 53 mm, 2 x 1/4"; 2/Pk
Representative image only.
Cole-Parmer Item # WZ-06060-29
Aseptic handling solutions for high-purity or biologically sensitive fluids
  • Eliminate pouring of high-value products through this leakproof aseptic system
  • Molded-in grips allow easy placement and removal
  • Multiple cap sizes to fit carboy threads from most manufacturers
  • Choose caps with solid silicone insert or ported with silicone tubing
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPP
  • Cap / Stopper Size53B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID2 x 1/4\" ID tubing (12\" L)
  • DescriptionEasy-Open Carboy Cap, 53B, 2 x 1/4\" Tubing; 1 Ea
Key Features
  • Eliminate pouring of high-value products through this leakproof aseptic system
  • Molded-in grips allow easy placement and removal
  • Multiple cap sizes to fit carboy threads from most manufacturers
  • Choose caps with solid silicone insert or ported with silicone tubing
More About this Item
Suggested autoclaving at 15 psi (1.03 bar) and 121˚C (250˚F) for 20 minutes.