Foxx Life Sciences 205-6103-CP Carboy Cap, 83 mm, 2 x 1/4" Hose Barb, with Tubing
Representative image only.
Mfr # 205-6103-RLS – Item # WZ-06059-33
Eliminate pouring expensive solutions with these aseptic caps
  • Retrofit to your system—choose from common carboy cap sizes
  • Designed to accept connectors and air vent filters
  • Polypropylene and high-purity silicone materials may be autoclaved
£98.11GBP / Each
£117.73GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPolypropylene and silicone
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID2 x 1/4\" ID
  • DescriptionFilling and Venting Carboy Cap, 83B, 2 x 1/4\" ID; 1/Ea
Key Features
  • Retrofit to your system—choose from common carboy cap sizes
  • Designed to accept connectors and air vent filters
  • Polypropylene and high-purity silicone materials may be autoclaved
More About this Item
Foxx Life Sciences™ VersaCap® Filling and Venting closures are aseptic handling solutions for pharmaceutical, biotechnology, and laboratory liquid transfer applications. Eliminate pouring of high valued products through this leak proof aseptic system.