Cole-Parmer 155162-43 Double open end long-life carbon block cartridge; 0.5 µm, 10"L
Representative image only.
Cole-Parmer Item # WZ-01508-93
£20.02GBP / Each
£24.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaActivated Carbon
  • Micron Rating0.5
  • Flow Rate1 GPM at 3.7 psid
  • Filter Length (in)10
  • Max Temperature (° F)180
  • Max Temperature (° C)82.2
  • DescriptionDouble open end long-life carbon block cartridge; 0.5 µm, 10"L
More About this Item
Long-Life Carbon Block Cartridges have a polyolefin-bonded powdered activated carbon (PAC) filter. Surface area of 2.2 million sq ft. Removes tastes, odors, chlorine, and sediment. Contains approximately 12 oz of carbon. PP end caps and outer wraps. Maximum temperature is 100°F (38°C); maximum differential pressure is 150 psid. For optimal performance, change cartridges at 25 psid.
** Not approved for residential use in the State of California