Cole-Parmer 155071-43 Double open end polypropylene string-wound cartridge; 5 µm, 10"L
Representative image only.
Cole-Parmer Item # WZ-01513-00
String-wind pattern has a lower presure drop for higher flow
PRODUCT OPTIONS
£9.90GBP / Each
£11.88GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating5
  • Flow Rate7 GPM
  • Filter Length (in)10
  • DescriptionDouble open end polypropylene string-wound cartridge; 5 µm, 10"L
More About this Item
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene string is resistant to chemical and bacterial attack. Maximum temperature is 150°F (66°C).
Note: For optimal performance, change cartridge at 25 psid.