Cole-Parmer 0/120 Percent Saturation Calcium Chloride Salometer
Representative image only.
Cole-Parmer Item # WZ-08299-21
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
  • 5 year warranty
PRODUCT OPTIONS
£37.85GBP / Each
£45.42GBP (incl VAT)
£141.67 - £200.21GBP / Each
£170.00 - £240.25GBP (incl VAT)
for an additional
£141.67GBP / Each
£170.00GBP (incl VAT)
for an additional
£200.21GBP / Each
£240.25GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Hydrometer FormPlain
  • Length (\")12
  • Division0.01
  • Accuracy±1.0%
  • Length (mm)300
  • Description 0/120 Percent Saturation Calcium Chloride Salometer
  • Warranty5 years
5 Year Warranty
Key Features
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
  • 5 year warranty
More About this Item
Ideal for determining the percentage of saturation, specific gravity or freezing point of CaCl2. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Equivalent to H-B Instrument 618074500.