This item is now discontinued. Please browse similar items
Chromatography Research Supplies Puresep-P 230640 Puresep-P Septa, 3/8" (9.5 mm); 50/Pk
Representative image only.
Chromatography Research Supplies Mfr # 230640 – Item # WZ-01994-01
Designed for enhanced bond and durability
  • Constructed of silicone rubber with a stable layer of polyimide film
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.375
  • DescriptionSepta, 3/8\" (9.5 mm); 50/Pack
Key Features
  • Constructed of silicone rubber with a stable layer of polyimide film
More About this Item
PURESEP-P™ septa are made of silicone rubber with a stable layer of polyimide film; withstand temperatures to 572°F (300°C).