This item is now discontinued. Please browse similar items

Chromaflex Standard Chromatography Column, 2172 mL, 120 cm L x 4.8 cm ID

Representative image only.
Chromaflex Mfr # 420830-1220 – Item # WZ-34696-22
Unique o-ring design provides a solvent resistant, leak-free seal
  • Constructed of borosilicate glass 3.3, PTFE, and HDPE for exceptional chemical resistance

SPECS & DESCRIPTION

Specifications & Description

  • SPE Cartridge Length (mm)1200
  • SPE Cartridge Length (cm)120
  • SPE Cartridge Diameter (cm)4.8
  • Tubing Outside Diameter (OD) (in)0.125
  • DescriptionStandard Chromatography Column, 2172 mL, 120 cm L x 4.8 cm ID

Key Features

  • Constructed of borosilicate glass 3.3, PTFE, and HDPE for exceptional chemical resistance

More About this Item

CHROMAFLEX® chromatography columns feature uniquely designed PTFE shielded o-rings to form a solvent resistant, leak-free seal. Each end fitting has a thin PTFE layer that is curled over the o-ring. Columns can be used with all aqueous buffers without modification and with organic solvents when the optional PTFE bed supports are installed in the end fittings and flow adapters. Columns also feature borosilicate glass barrel construction with PTFE end fittings and HDPE bed supports. Recommended sterilization is with ethylene oxide, 2N NaOH or 100% ethanol (autoclaving is not recommended).

Choose from standard or jacketed columns. Jacketed columns feature a water jacket around the barrel to provide thermal control of temperature-sensitive chromatographic separations. The jacket also acts as a safety shield.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat