Chemical Resistant Pan - 25-1/4" X 19-1/4", 1 Ea

Representative image only.
Item # WZ-06715-86
Resistant to mild acids, alkaline solutions and many reagents.
  • Withstand temperatures from –40 to 250°F (–40 to 121°C)
  • Contain spills with straight edge pans

PRODUCT OPTIONS

£70.77GBP / Each
£84.92GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • ColorWhite
  • MaterialPolyester
  • Overall Length (in)25 1/4
  • Overall Length (cm)64.1
  • Overall Width (in)19 1/4
  • Overall Width (cm)48.9
  • Overall Height (in)1 1/8
  • Overall Height (cm)2.9
  • DescriptionChemical Resistant Pan - 25-1/4\" X 19-1/4\", 1 Ea

Key Features

  • Withstand temperatures from –40 to 250°F (–40 to 121°C)
  • Contain spills with straight edge pans

More About this Item

Constructed of Thermoset polyester resin, these white trays are ideal for carrying, storing, and cleaning of lab product. Trays withstands temperatures from 40°C to 121° (-40°F to 250°F). Pans with straight edges are ideal for spill containment.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat