This item is now discontinued. We recommend the following as suggested replacements: WZ-06925-00
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 15 gallon
Representative image only.
Chem-Tainer Mfr # TC1530BP-FA0100-INSTALLED – Item # WZ-06929-04
Tanks give you excellent chemical compatibility
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Volume (Gal)15
  • Volume (Liters)57
  • Height (in)30
  • Height (cm)76.2
  • Outside Diameter (OD) (in)15
  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • DescriptionConical-Bottom Open-Top Tank with Stand, PP, 15 gallon
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.