This item is now discontinued. Please browse similar items
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 100 gallon
Representative image only.
Chem-Tainer Mfr # TC3048BP-FA0150-INSTALLED – Item # WZ-06929-12
Tanks give you excellent chemical compatibility
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Volume (Gal)100
  • Volume (Liters)379
  • Height (in)48
  • Height (cm)121.92
  • Outside Diameter (OD) (in)30
  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • DescriptionConical-Bottom Open-Top Tank with Stand, PP, 100 gallon
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.