CELLTREAT Scientific Products Cap for Erlenmeyer/Fernbach Flasks, Sterile, Polypropylene, 70 mm, Vented, 0.22 µm; 24/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229895 – Item # WZ-01931-83
Cell culture caps—ideal for aseptic biological reactions
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Autoclavable
  • Over-sized vent area in cap is ideal for improved gas exchange
  • Individually packaged to reduce cross contamination
PRODUCT OPTIONS
£168.35GBP / Case of 24
£202.02GBP (incl VAT)
Often Bought With
CELLTREAT Vented Sterile Culture Flasks
£278.56 - £328.67GBP / Case of 6
£334.27 - £394.40GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • AutoclavableYes
  • Cap IncludedCap Only
  • Cap StyleVented
  • Cap MaterialPolypropylene
  • PackagingIndividually packaged
  • Special FeaturesFor use with CELLTREAT erlenmeyer and fernbach flasks
  • DescriptionCap for Erlenmeyer/Fernbach Flasks, Sterile, Polypropylene, 70 mm, Vented, 0.22 µm; 24/Cs
Bio DNase-FreeNonpyrogenicRNase-FreeSterile
Key Features
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Autoclavable
  • Over-sized vent area in cap is ideal for improved gas exchange
  • Individually packaged to reduce cross contamination
More About this Item
These convenient replacement caps for your tubes, bottles, and flask are sterilized by gamma irradiation and ideal for cell culture applications. Choose from solid or vented caps. Solid caps provide a leakproof seal to keep samples secure. Vented caps feature a 0.22 µm hydrophobic membrane allowing for continuous gas exchange.
Choose the cap the is suitable for use with your CELLTREAT® Scientific Products cell culture product. Roller bottles, flasks, and centrifuge tubes sold separately.