CELLTREAT Scientific Products Cap for Bio-Reaction Centrifuge Tube, Sterile, HDPE, for 15 mL Tube, Vented, 0.22 µm; 100/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229473 – Item # WZ-06336-42
Cell culture caps—ideal for aseptic biological reactions
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Vented cap for 15 mL Bioreaction Tubes
  • Packaged 25 caps per resealable bag
PRODUCT OPTIONS
£136.03GBP / Case of 100
£163.24GBP (incl VAT)
Often Bought With
CELLTREAT Scientific Products Bio-Reaction Centrifuge Tubes
£146.44 - £471.20GBP / Case of 100
£175.73 - £565.44GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Cap IncludedCap Only
  • Cap StyleVented
  • Cap MaterialHDPE
  • Packaging25/Bag, 4 Bags/Case
  • Special FeaturesFor use with CELLTREAT 15 mL bio-reaction centrifuge tubes
  • DescriptionCap for Bio-Reaction Centrifuge Tube, Sterile, HDPE, for 15 mL Tube, Vented, 0.22 µm; 100/Cs
Bio DNase-FreeNonpyrogenicRNase-FreeSterile
Key Features
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Vented cap for 15 mL Bioreaction Tubes
  • Packaged 25 caps per resealable bag
More About this Item
These convenient replacement caps for your tubes, bottles, and flask are sterilized by gamma irradiation and ideal for cell culture applications. Choose from solid or vented caps. Solid caps provide a leakproof seal to keep samples secure. Vented caps feature a 0.22 µm hydrophobic membrane allowing for continuous gas exchange.
Choose the cap the is suitable for use with your CELLTREAT® Scientific Products cell culture product. Roller bottles, flasks, and centrifuge tubes sold separately.