CELLTREAT Scientific Products Reagent Reservoir, Polystyrene, Sterile, 50 mL, Individually Wrapped; 80/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229290 – Item # WZ-34567-58
Ideal for repetitive liquid transfer with multichannel pipettors
  • Reservoirs are presterilized by gamma irradiation
  • V-shaped angled design promotes funneling of liquid to maximize retrieval and reduce waste
  • Constructed of durable white polystyrene for enhanced solution visibility
  • Individually wrapped reservoirs come in a case of 80
PRODUCT OPTIONS
£160.67GBP / Case of 80
£192.80GBP (incl VAT)
Sugggested alternative
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)50
  • Wetted MatterialPolystyrene
  • Color CodeWhite
  • Package OfIndividually Wrapped Case of 80
  • DescriptionReagent Reservoir, Polystyrene, Sterile, 50 mL, Individually Wrapped; 80/Cs
Sterile
Key Features
  • Reservoirs are presterilized by gamma irradiation
  • V-shaped angled design promotes funneling of liquid to maximize retrieval and reduce waste
  • Constructed of durable white polystyrene for enhanced solution visibility
  • Individually wrapped reservoirs come in a case of 80
More About this Item
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. Sterile reservoirs are sterilized by gamma irradiation and come individually wrapped or bulk packed.