CELLTREAT Scientific Products 229226B Short-Length Serological Pipette, 25mL, Indv. Wrap, Paper/Plastic, Sterile; 200/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229226B – Item # WZ-34567-41
Bold, black ink indicate positive and negative graduations
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Ideal for compact workspaces and designed with ergonomics in mind
PRODUCT OPTIONS
£97.74GBP / Case of 200
£117.29GBP (incl VAT)
Sugggested alternative
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)25
  • Graduation Interval0.5 mL
  • Increments (mL)0.5
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodeRed
  • Filter TypeMaterial plug
  • Package Of25 individually wrapped per bag
  • DescriptionShort-Length Serological Pipet, 25mL, Indv. Wrap, Paper/Plastic, Sterile; 200/Cs
Sterile
Key Features
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Ideal for compact workspaces and designed with ergonomics in mind
More About this Item
These sterile serological pipettes feature bold black positive and negative graduations making them easy to read while flling or dispensing. Color coded stripes and packaging enable quick volume identification.